Learn More
Kimble Chase™ Borosilicate Glass Plain Chromatography Column
Product Code. 11867453
Change view
Click to view available options
Column Capacity:
15 mL
25 mL
340 mL
440 mL
567 mL
589 mL
900 mL
965 mL
981 mL
1608 mL
1929 mL
2149 mL
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity unitSize
11867453 15 mL Each
11877483 1608 mL Each
11887483 1929 mL Each
11837493 2149 mL Each
11877453 25 mL Each
11827483 340 mL Each
11857473 440 mL Each
11837483 567 mL Each
13446349 900 mL Each
11857483 981 mL Each
11867483 965 mL Each
11847483 589 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11867453 Supplier Kimble Chase™ Supplier No. 4205300212
 Restricted Item
Estimated Shipment: 15-01-2027
Specifications
USP Type Type I
Column Capacity 15 mL
Length (Metric) 200 mm
Material Borosilicate Glass
Includes With PTFE stopcock plug
Quantity 1 Each
For Use With (Application) Ion exchange chromatography
Type Plain Chromatography Column
Diameter (Metric) Inner 11 mm