Learn More
Scientific Glass Labs™ Borosilicate Glass Spheres
Product Code. 12848353
Change view
Click to view available options
Diameter (Metric):
3 mm
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12848353 3 mm 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12848353 Supplier Scientific Glass Labs™ Supplier No. GS/3
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with distillation columns. Borosilicate glass construction.
  • Borosilicate glass construction
  • Supplied in 500 gram bag ready for use
  • For use with distillation columns
Specifications
Type Beads
Material Borosilicate Glass
Diameter (Metric) 3 mm
For Use With (Equipment) Distillation Columns
Quantity 500 g/bag