Learn More
Hamilton™ 7000 Series Knurled Hub Syringes
Click to view available options
Needle Gauge:
23 G
25 G
Unit Size:
Each
Volume (Metric):
1 μL
Description
- The only zero dead volume syringe available
- Fine gauge tungsten plunger wire extends to the tip of the needle
- Ensures that all solvent is expelled
Specifications
Specifications
| Material | Borosilicate Glass, Stainless Steel |
| Needle Gauge | 23 G |
| Product Type | Syringe |
| Quantity | 1/Ea. |
| Needle Point Style | 3 |