missing translation for 'onlineSavingsMsg'
Learn More

Boscalid, TRC

Product Code. 30008611
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008611 10 g 10g
30012317 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008611 Supplier Toronto Research Chemicals Supplier No. TRCB67585010G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188425-85-6
Molecular Formula C18 H12 Cl2 N2 O
Molecular Weight (g/mol) 343.21
Synonym 2-Chloro-N-(4'-chloro[1,1'-biphenyl]-2-yl)-3-pyridinecarboxamide, 2-Chloro-N-(4'-chlorobiphenyl-2-yl)nicotinamide, BAS 510F, Boscalid, Cantus, Emerald, Emerald (fungicide), Endura, F 510, Filan, Nicobifen
IUPAC Name 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide
SMILES Clc1ccc(cc1)c2ccccc2NC(=O)c3cccnc3Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C18H12Cl2N2O/c19-13-9-7-12(8-10-13)14-4-1-2-6-16(14)22-18(23)15-5-3-11-21-17(15)20/h1-11H,(H,22,23)
Formula Weight 342.0327
Chemical Name or Material Boscalid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.