missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Brain Extract

Product Code. 16339865 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
2.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16339865 2.5 mL 2.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16339865 Supplier MP Biomedicals™ Supplier No. 092850550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filtered Bovine Brain Extract used for various endothelial cell type culture.

  • Bovine Brain Extract (BBE) is made from fresh, unfrozen pituitary glands from young, healthy cattle less than 30 months old. They are born, raised and slaughtered in New Zealand from a USDA approved source. New Zealand cattle are internationally recognized for being free from bovine spongiform encephalopathy (BSE, mad-cow disease) and have no history of blue tongue disease.
  • This Bovine Brain Extract (BBE) is formulated to be used at 0.1-0.2 % in the final serum-free medium. Studies have shown this level of BBE at the standard 7.5-15 micrograms/ mL will fully replace endothelial cell growth supplement (ECGS). BBE allows culture of these cells without feeder layers.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C, do not thaw and refreeze
Form Liquid
Sterility Sterile
For Use With (Application) Cell Culture
Quantity 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.