Learn More
Abcam Bovine Heart Mitochondria, 10 mg
Product Code. 32142131 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32142131 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32142131 Supplier Abcam Supplier No. ab11033810mg
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Bovine Heart cell/tissue lysate.
  • Species: Bovine
  • Tissue: Heart
  • Form: Liquid
Specifications
For Use With (Application) Western Blot, SDS-PAGE, Immunoprecipitation