missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BPI Antibody (SR3018), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00€ - 382.00€
Specifications
| Antigen | BPI |
|---|---|
| Clone | SR3018 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041623
|
Novus Biologicals™
NBP3-27733-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041682
|
Novus Biologicals™
NBP3-27733-50ul |
50 μL |
316.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BPI Monoclonal antibody specifically detects BPI in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| BPI | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 671 | |
| IgG | |
| Affinity purified |
| SR3018 | |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, CAP 57 | |
| A synthesized peptide derived from human BPI (Uniprot #: P17213) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title