Learn More
Trajan™ BPX5 GC Capillary Columns
Product Code. 10616021
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Length (Metric):
6 m
10 m
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.25 μm
0.5 μm
1 μm
missing translation for 'unitSize':
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness missing translation for 'unitSize'
10616021 30 m 0.32 mm 0.25 μm Each
15218653 6 m 0.32 mm 1 μm Each
10567342 12 m 0.22 mm 0.25 μm Each
10442341 25 m 0.32 mm 0.25 μm Each
10519452 12 m 0.53 mm 0.25 μm Each
12684945 60 m 0.25 mm 0.25 μm Each
16662612 60 m 0.32 mm 1 μm Each
16642612 30 m 0.53 mm 1 μm Each
16622612 25 m 0.53 mm 1 μm Each
10320941 50 m 0.22 mm 0.25 μm Each
10269190 25 m 0.53 mm 0.25 μm Each
15124079 30 m 0.53 mm 0.5 μm Each
10430081 25 m 0.32 mm 0.5 μm Each
10268810 25 m 0.22 mm 0.25 μm Each
10451031 25 m 0.15 mm 0.25 μm Each
10791412 30 m 0.22 mm 0.25 μm Each
10208670 60 m 0.32 mm 0.25 μm Each
10567532 12 m 0.32 mm 0.25 μm Each
10025450 12 m 0.53 mm 1 μm Each
10300711 12 m 0.32 mm 1 μm Each
15288643 15 m 0.25 mm 1 μm Each
10177200 15 m 0.32 mm 0.25 μm Each
12905561 15 m 0.25 mm 0.25 μm Each
10762562 50 m 0.32 mm 0.5 μm Each
15298643 30 m 0.25 mm 1 μm Each
10661813 10 m 0.1 mm 0.1 μm Each
10055400 50 m 0.32 mm 1 μm Each
15248643 30 m 0.15 mm 0.15 μm Each
15238643 30 m 0.25 mm 0.1 μm Each
15228663 15 m 0.25 mm 0.1 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 missing translation for 'options'
Product Code. 10616021 missing translation for 'mfr' Trajan™ missing translation for 'supplierNo' 054145
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
For Use With (Application) Amines Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines
Film Thickness 0.25 μm
Length (Metric) 30 m
Packing Material BPX5
Polarity Non polar
Coating Fused Silica
USP Type G27, G36, G41
Stationary Phase 5% Phenyl Polysilphenylene Siloxane
Diameter (Metric) Inner 0.32 mm
Functionality/Interaction Type Pi Bonding with the Aromatic Phenyl Groups
Max. Temperature (Metric) 370°C
Temperature Range (Metric) -40°C to +360/370°C