missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Brachyury Monoclonal antibody specifically detects Brachyury in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Brachyury |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | TBXT/7711R |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT |
| Host Species | Rabbit |
| Immunogen | Recombinant fragment (around aa 235-435) of human Brachyury protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?