missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP™ 71 PermaSleeve™ Wire Marker Sleeves

Product Code. 11548594
Change view
Click to view available options
Diameter (Metric):
1.17 to 2.79 mm
1.57 to 3.81 mm
2.39 to 5.46 mm
3.2 to 8.13 mm
4.75 to 11.43 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11548594 1.17 to 2.79 mm Each
11558594 1.57 to 3.81 mm Each
11568594 2.39 to 5.46 mm Each
11578594 3.2 to 8.13 mm Each
11588594 4.75 to 11.43 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11548594 Supplier Brady™ Supplier No. 115118

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Fits snuggly around wire for permanent identification and protection

  • Has 3:1 shrink ratio 
  • Heat shrinkable 
  • Fade resistant
  • Flame retardant
  • Made of B-342 Permasleeve PS polyolefin material

Specifications

Certifications/Compliance SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202, RoHS Compliant, UL Recognized; Tested per: ASTM E162 and ASTM E662
Description Bulk BMP71 PermaSleeve Wire Marker Sleeves
Material Polyolefin (B-342)
Product Type Wire Marker Sleeve
For Use With (Application) Wire and cable marking, Panel identification
For Use With (Equipment) 22 to 16 wire ga
Quantity 1
Length (Metric) 15.24 m
Diameter (English) 0.046 to 0.110 in.
Diameter (Metric) 1.17 to 2.79 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.