Learn More
Brady™ BradyJet J4000 Color Label Printer
Product Code. 17459212
Change view
Click to view available options
Color:
Blue, Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17459212 Blue, Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17459212 Supplier Brady™ Supplier No. 152711
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
High-impact, precision full color that lasts. Create the compliant, long-lasting labels, signs and tags you need with the BradyJet J4000 inkjet printer.
  • This industry inkjet workhorse creates up to 5,000 impactful full-color labels per day up to 203.20 mm wide on continuous or die-cut material
  • Ideal for a wide variety of applications in safety, including Lean / 5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings
  • Contains the color options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels
  • Streamline storage by only needing to stock white label material and print only what you need, when you need it, in photo-quality full colour every time
  • Easily print PDFs or from third-party software packages with the optimised Windows driver
  • Add Brady Workstation Safety and Facility ID Software Suite (J4000-BWSSFID) or Lab Suite (J4000-BWSLAB) to your solution and make label creation even easier
Specifications
Product Type Printer
Color Blue, Gray
Display Type LED
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Inkjet
Length (English) 9.1 in.
Width (English) 17.25 in.
Length (Metric) 231.14 mm
Width (Metric) 438.15 mm
Depth (English) 17.25 in.
Depth (Metric) 438.15 mm
Color Capability Full Color
Description EU Plug Type
Includes 1 x BradyJet J4000 Printer, 1 x Power Cord, 1 x CMY Ink Cartridge with built in Printhead, 1 x Pack of 2 Ink Cartridge Storage Caps, 1 x Media starter roll 101.60 mm wide B-2595 Vinyl, 1 x USB Cord, 1 x USB Flash Drive with product information, 1 x Brady Workstation Basic Suite
Model J4000
Print Speed 114.30 mm/s
Resolution 4800 dpi
Connector Types USB, Ethernet
Weight (English) 36 lb.
Weight (Metric) 16 kg
Certifications/Compliance CE, C-UL US, FCC Class A, RoHS directive 2011/65/EU, UL Listed for US
For Use With (Application) Facility identification, General identification, Hazard communication identification, Lean and 5S labelling, Pipe and valve marking, Warehouse marking
Quantity Each
Warranty 1 Year
Power Supply Details AC Adapter
Voltage 100/240 V
Keyboard Type None
Alternate Part Number J4000-EU