missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyJet™ J7300 Color Label Printer EU and BWS Suite

Product Code. 30058013
Change view
Click to view available options
Color:
Blue, Gray
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30058013 Blue, Gray 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30058013 Supplier Brady™ Supplier No. 322660
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Unleash your true colors with the BradyJet™ J7300 Color Label Printer. Enjoy fast, industrial-grade full color for variable print jobs from the production line to the lab.

  • Ideal for industrial labelling that changes frequently, from regulatory signage and Lean visuals to barcoding, lab ID, GHS labelling and UL/component ID
  • LabelSense™ technology enables printer, materials and inks to talk to each other for automatic part recognition, setup and adjustments
  • Dual CMY and true black (K) ink cartridges deliver vibrant colors, deep black, dark tones, readable barcodes and sharp text job after job
  • Prints smear-free inks on labels that resist water, chemicals and abrasions for up to two-year outdoor durability (even longer indoors)
  • Print Utility dashboard helps you plan, budget and troubleshoot with status alerts, a job cost calculator, remote monitoring and more
  • Exterior LED lighting and clear cover communicate real-time printer status
  • Ships with a license for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
  • Optimized Windows™ driver lets you connect to third-party software and print PDFs

Specifications

Product Type Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Inkjet
Length (English) 9.09 in.
Width (English) 14.8 in.
Length (Metric) 230.9 mm
Width (Metric) 376.3 mm
Depth (English) 20.3 in.
Depth (Metric) 515.7 mm
Color Capability Full Color
Description BradyJet J7300 Color Label Printer EU + Brady Workstation Suite
Display Size 71.12 mm
Includes BradyJet J7300 Printer, CMY Ink Cartridge with built-in Printhead, a Black Ink Cartridge with Built-in Printhead, a License for Brady Workstation Software Suites, a J-Align Roll, a Media Starter B-2595 Vinyl Roll, an AC Adapter, a Power Cord, a USB Cable and a USB Flash Drive with Product Information.
Languages Bahasa (Indonesian), Bahasa Malay, Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Greek, Hungarian, Italian, Japanese, Korean, Norwegian (Bokmål), Polish, Portuguese, Punjabi, Slovak, Slovenian, Spanish (universal), Swedish, Thai, Turkish, Vietnamese
Memory 32 GB, expandable through USB
Model J7300
Print Speed 3.05 mm/s
Resolution 4800 dpi
Software Type Brady LINK260;Brady Workstation;Windows based driver for 3rd party software use
Connector Types Cabled, Ethernet, USB-C, USB Host
Operating System Windows 10
Weight (English) 36 lb
Weight (Metric) 16 kg
Certifications/Compliance CE, C-UL US, FCC Class A, RoHS directive 2011/65/EU, UL Listed for US
For Use With (Application) Asset tracking, Barcode labelling, Component and equipment labelling, Exit and egress marking, Facility identification, General identification, Hazard communication identification, Healthcare and Hospital labelling, Inspection tagging, Inventory and inspection labelling, Laboratory labelling, Lean and 5S labelling, Lockout tagout, Marine labelling, Pipe and valve marking, Safety identification, Warehouse marking
Quantity 1
Warranty 2 Years
Amperage 100/240 V
Power Supply Details AC Adapter
Voltage 100/240 V
Keyboard Type None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.