Learn More
Brady™ BradyPrinter i7100 with Peel Function and Vial Label Applicator with BWS Suite
Product Code. 17107031 Shop All Brady Products
Change view
Click to view available options
Resolution:
300 dpi
600 dpi
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Resolution Unit Size
17107031 600 dpi 1 set
17197021 300 dpi 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17107031 Supplier Brady™ Supplier No. 307659
  • ONLINE EXCLUSIVE PRICE
    7340.00€
    6633.76€ / 1 sets
    Save 706.24€
    9% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing.
  • The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for customers requiring reliable high volume printing.
  • Automate vial tube identification with the Vial Label Applicator that prints and applies labels on vial tubes in less than 2 seconds.
  • The printer comes with the Brady Workstation Software suite enabling you to immediately start designing professional labels.
  • This suite combines frequently used software apps to create laboratory identification labels.
Specifications
Product Type Printer
Display Type Color Touchscreen
For Use With (Equipment) THT Series Printers
Print Technology Thermal Transfer
Width (English) 9.92 in.
Width (Metric) 252 mm
Height (English) 11.34 in.
Height (Metric) 288 mm
Depth (English) 20.47 in.
Depth (Metric) 520.00 mm
Dimensions (L x W x H) 520.00 x 252.00 x 288.00 mm
Color Capability Monocolour
Includes Vial label applicator; power cord; 2 empty ribbon cores; USB cable; CD with driver, user manual, the configuration/settings manual; printed user manual; Brady Workstation Software suites.
Languages Arabic, Bulgarian, Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Macedonian, Norwegian, Persian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
Memory 256 MB RAM, 50 MB IFFS Flash, SD card up to 512 GB
Print Speed 150.00 mm/sec.
Resolution 600 dpi
Software Type Brady Workstation, Windows based driver for 3rd party software use
Connector Types Ethernet;I/O connection;Bluetooth (optional);Wi-Fi (optional);Serial;USB
Operating System Windows 7;Windows 8;Windows 10;Windows Server 2012;Windows Server 2012 R2;Windows Server 2016
Weight (English) 26.46 lb.
Weight (Metric) 12 kg
Certifications/Compliance CE;FCC class A;FCC class CB;CCC;UL
For Use With (Application) Labeling
Quantity 1 Ea.
Warranty 2 Years
Power Supply Details AC Adaptor
Voltage 110 V
Alternate Part Number I7100600P-EUBWSVLA
Plug Type EU