Description
- Easy-to-follow procedures are outlined step-by-step and all calculations are performed automatically by the software.
- Weighing values may be entered manually into the software or imported directly from the balance via serial port.
- Generates reports to document the calibration testing in accordance to ISO 8655, ISO 4787 etc.
- All test records can be saved in a database for subsequent use and tracking of instrument performance.
- Simplifies data entry
- Organizes all test data
- Includes SOPs
- Clearly arranged test record
Specifications
Specifications
| Product Type | Calibration Software |
| For Use With (Application) | Calibration of liquid handling to ISO 9001 and GLP standards |
| Includes | CD Rom in English, French, German, Portuguese,and Spanish |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?