missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ plus, 96-well Flat Bottom Microplates, PS

Product Code. 15674017 Shop All Brand Products
Change view
Click to view available options
Packaging:
1/Pk., 50/Cs.
5/Pk.
Color:
Black, Clear Bottom
Clear
White, Clear Bottom, Clear Bottom
Unit Size:
Pack of 5
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Color Unit Size
15674017 5/Pk. White, Clear Bottom, Clear Bottom Pack of 5
10564584 1/Pk., 50/Cs. White, Clear Bottom, Clear Bottom Pack of 50
10627713 - Clear Pack of 50
10793074 - Black, Clear Bottom Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15674017 Supplier BRAND™ Supplier No. 782034
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile 96-well microplates with cellGrade™plus surface treatment for optimal cell growth. Features protein-like surface for fast cell attachment and better yield. Ideal for cultivating sensitive or fastidious cell lines, even in reduced-serum media

  • Hydrophilic surface with freely-accessible amino, carboxyl and hydroxyl groups
  • Protein-like composition allows for direct cellular attachment
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • BIO-CERT™ CELL CULTURE QUALITY: free of pyrogens, endotoxins, human DNA, RNase, and DNase
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ISO 11137, DIN EN ISO 10993
Format Standard
Lid With Lid
Array 8 x 12
Packaging 5/Pk.
Color White, Clear Bottom, Clear Bottom
Product Type 96-well plate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade Plus
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cells in reduced serum media and fastidious cell lines
No. per Pack 5/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.