missing translation for 'onlineSavingsMsg'
Learn More

BRD 7389, Tocris Bioscience™

Product Code. 30001432
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001432 10 mg 10mg
18094792 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001432 Supplier Tocris Bioscience Supplier No. 4037/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ribosomal S6 kinase inhibitor

Ribosomal S6 kinase (RSK) inhibitor (IC50 values are 1.2, 1.5 and 2.4 μM for RSK3, RSK1 and RSK2 respectively). Upregulates insulin expression in terminally differentiated pancreatic α-cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376382-11-5
Molecular Formula C24H18N2O2
Molecular Weight (g/mol) 366.42
InChI Key XASCINRGTHLHGM-UHFFFAOYSA-N
Synonym 1-2-phenylethyl amino-3h-naphtho 1,2,3-de quinoline-2,7-dione, 1-phenethylamino-3h-naphtho 1,2,3-de quinoline-2,7-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2 7 ,3,5,9 17 ,10,12-heptaene-8,15-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2 7 ,3,5,9,11,13 17-heptaene-8,15-dione, 16-2-phenylethyl amino-14-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1 16 ,2,4,6,9 17 ,10,12-heptaene-8,15-dione
PubChem CID 1080352
SMILES C1=CC=C(C=C1)CCNC2=C3C4=CC=CC=C4C(=O)C5=C3C(=CC=C5)NC2=O

Specifications

Quantity 10 mg
Formula Weight 366.41
Percent Purity >97%
Chemical Name or Material BRD 7389
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.