missing translation for 'onlineSavingsMsg'
Learn More

Breast Adenocaricinoma (MCF-7) Total RNA

Product Code. 31782135 Shop All Applied Biosystems Products
This item is not returnable. View return policy
Product Code. 31782135 Supplier ABI Applied Biosystems Supplier No. QS0600

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Species Human
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Total RNA
Quantity 10 μg
Sample Source Tumor or Diseased Tissue, Cell Line
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.