missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Brefeldin A

Product Code. 17130162
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130162 25 mg 25mg
17120162 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17130162 Supplier STEMCELL Technologies Supplier No. 73014
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Protein trafficking inhibitor; Inhibits Sec7-containing guanine-exchange factor (GEF)

  • Brefeldin A is a fungal lactone antibiotic, produced by many species, including Eupenicillium brefeldianum (Klausner et al.) It reversibly interferes with protein trafficking and secretion mediated by the Golgi apparatus and endoplasmic reticulum by indirect inhibition of ADP-ribosylation factor (ARF; Klausner et al.; Helms & Rothman; Robinson et al.; Morinaga et al.; Moss & Vaughan; Nebenführ et al.; Ktistakis et al.) Brefeldin A binds to Sec7-containing guanine-exchange factor (GEF) at the ARF-GDP-Sec7 interface, preventing the conformational change required to release GDP and activate ARF (Mossessova et al.)

Specifications

CAS 20350-15-6
Synonym Ascotoxin; BFA; Cyanein; Decumbin; Nectrolide; NSC 56310; NSC 89671; NSC 107456; NSC 244390; Synergisidin
Quantity 25 mg
Molecular Formula C16H24O4
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.