missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Brefeldin A

Product Code. 18055762
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055762 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055762 Supplier Tocris Bioscience™ Supplier No. 1231/5
  • ONLINE EXCLUSIVE PRICE
    245.00€
    231.00€ / 5mg
    Save 14.00€
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Disrupts protein translocation to Golgi

Reversible inhibitor of protein translocation from the endoplasmic reticulum (ER) to the Golgi apparatus. Blocks binding of ADP-ribosylation factor (ARF1) to the Golgi apparatus and inhibits GDP-GTP exchange. Can be used to induce autophagy in mammalian cells. Also enhances CRISPR-mediated homology-directed repair (HDR) efficiency ∼2-fold when applied at 100 nM, in human induced pluripotent stem cells (iPSCs). Antifungal.
TRUSTED_SUSTAINABILITY

Specifications

CAS 20350-15-6
Quantity 5 mg
Molecular Formula C16H24O4
Formula Weight 280.36
Purity >98%
Molecular Weight (g/mol) 280.36
Chemical Name or Material 1,6,7,8,9,11 Aβ,12,13,14,14 Aα-Decahydro-1β,13α-dihydroxy-6β-methyl-4 H-cyclopent[f]oxacyclotridecin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.