Learn More
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35
Product Code. 10124810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
HDPE plastic bottle
Plastic drum
Unit Size:
1kg
250g
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124810 250 g HDPE plastic bottle 250g
10043100 500 g HDPE plastic bottle 500g
10226430 1 kg HDPE plastic bottle 1kg
11956941 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10124810 Supplier Thermo Scientific Acros Supplier No. 228342500
  • ONLINE EXCLUSIVE PRICE
    45.10€
    38.65€ / 250g
    Save 6.45€
    14% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
missing translation for 'validMainframeMsg'
CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene(23)lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO
Specifications
Color White
Acid Value 5mg KOH/g max.
Physical Form Waxy Pellets
Boiling Point >100.0°C
Melting Point 33.0°C to 40.0°C
Quantity 250 g
Flash Point >149°C
Hydroxyl Value 40 to 60mg KOH/g
Infrared Spectrum Authentic
Packaging HDPE plastic bottle
Linear Formula C12H25(OCH2CH2)23OH
Water 1% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils
Formula Weight 1199.56
Chemical Name or Material Brij™ 35