missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brimonidine, 98%

Product Code. 15628745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15628745 1 g 1g
15638745 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15628745 Supplier Thermo Scientific Chemicals Supplier No. 461520010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 59803-98-4 | C11H10BrN5 | 292.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59803-98-4
Molecular Formula C11H10BrN5
Molecular Weight (g/mol) 292.14
MDL Number MFCD00153878
InChI Key XYLJNLCSTIOKRM-UHFFFAOYSA-N
Synonym brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
PubChem CID 2435
ChEBI CHEBI:3175
IUPAC Name 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
SMILES BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1

Specifica

Melting Point 235.0°C to 236.0°C
Ignition Residue 0.3% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 292.13
Percent Purity 98%
Chemical Name or Material Brimonidine

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.