missing translation for 'onlineSavingsMsg'
Learn More

Broflanilide, TRC

Product Code. 17994695
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994695 50 mg 50mg
17964965 25 mg 25mg
17963145 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994695 Supplier Toronto Research Chemicals Supplier No. TRCB67757050MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207727-04-5
Molecular Formula C25 H14 Br F11 N2 O2
Molecular Weight (g/mol) 663.28
Synonym 3-(Benzoylmethylamino)-N-[2-bromo-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]-6-(trifluoromethyl)phenyl]-2-fluorobenzamide, 2-Fluoro-3-(N-methylbenzamido)-N-[2-bromo-4-(heptafluoroisopropyl)-6-(trifluoromethyl)phenyl]benzamide, 3-[Benzoyl(methyl)amino]-N-[2-bromo-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]-6-(trifluoromethyl)phenyl]-2-fluoro-benzamide, Broflanilide, N-[2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)phenyl]-2-fluoro-3-(N-methylbenzamido)benzamide
IUPAC Name 3-[benzoyl(methyl)amino]-N-[2-bromo-4-(1,1,1,2,3,3,3-heptafluoropropan-2-yl)-6-(trifluoromethyl)phenyl]-2-fluorobenzamide
SMILES CN(C(=O)c1ccccc1)c2cccc(C(=O)Nc3c(Br)cc(cc3C(F)(F)F)C(F)(C(F)(F)F)C(F)(F)F)c2F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C25H14BrF11N2O2/c1-39(21(41)12-6-3-2-4-7-12)17-9-5-8-14(18(17)27)20(40)38-19-15(23(29,30)31)10-13(11-16(19)26)22(28,24(32,33)34)25(35,36)37/h2-11H,1H3,(H,38,40)
Formula Weight 662.0063
Chemical Name or Material Broflanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.