Learn More
Bromantane, TRC
Product Code. 30133406
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133406 5 g 5g
17942655 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133406 Supplier Toronto Research Chemicals Supplier No. TRCB6784505G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87913-26-6
Molecular Formula C16 H20 Br N
Molecular Weight (g/mol) 306.24
Synonym Tricyclo[3.3.1.13,7]decan-2-amine, N-(4-bromophenyl)-, N-(4-Bromophenyl)tricyclo[3.3.1.13,7]decan-2-amine, Bromantan, Bromantane
IUPAC Name N-(4-bromophenyl)adamantan-2-amine
SMILES Brc1ccc(NC2[C@@H]3C[C@H]4C[C@H](C[C@@H]2C4)C3)cc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C16H20BrN/c17-14-1-3-15(4-2-14)18-16-12-6-10-5-11(8-12)9-13(16)7-10/h1-4,10-13,16,18H,5-9H2/t10-,11+,12-,13+,16?
Formula Weight 305.0779
Percent Purity >95%
Chemical Name or Material Bromantan
Grade HPLC