missing translation for 'onlineSavingsMsg'
Learn More

Bromethalin, TRC

Product Code. 30012072
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012072 10 mg 10mg
30010437 100 mg 100mg
30011051 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012072 Supplier Toronto Research Chemicals Supplier No. TRCB63333010MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63333-35-7
Molecular Formula C14 H7 Br3 F3 N3 O4
Molecular Weight (g/mol) 577.93
Synonym N-Methyl-2,4-dinitro-N-(2,4,6-tribromophenyl)-6-(trifluoromethyl)benzenamine, Bromethalin, EL 614, Rampage, Rampage (rodenticide)
IUPAC Name N-methyl-2,4-dinitro-N-(2,4,6-tribromophenyl)-6-(trifluoromethyl)aniline
SMILES CN(c1c(Br)cc(Br)cc1Br)c2c(cc(cc2C(F)(F)F)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H7Br3F3N3O4/c1-21(13-9(16)2-6(15)3-10(13)17)12-8(14(18,19)20)4-7(22(24)25)5-11(12)23(26)27/h2-5H,1H3
Formula Weight 574.7939
Percent Purity >95%
Chemical Name or Material Bromethalin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.