missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoacetic Acid-d3, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Bromoacetic acid-d3 |
| CAS | 14341-48-1 |
| Molecular Formula | C2 D3 Br O2 |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)/i1D2/hD |
| SMILES | [2 H]OC(=O)C([2 H])([2 H])Br |
| IUPAC Name | deuterio 2-bromo-2,2-dideuterio-acetate |
| Molecular Weight (g/mol) | 141.97 |
| Formula Weight | 140.9505 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction