missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromobenzene-2,4,6-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | Bromobenzene-2,4,6-D3 |
| CAS | 13122-41-3 |
| Molecular Formula | C6 D3 H2 Br |
| Quantity | 0.5 g |
| Synonym | Benzene-1,3,5-d3, 2-bromo- (8 CI, 9 CI, ACI), 2-Bromobenzene-1,3,5-d3 (ACI), Benzene-1,3,5-d3, bromo- (6 CI, 7 CI), 2,4,6-d3-Bromobenzene, Bromobenzene-2,4,6-d3, Bromobenzene-2,4,6-D3 |
| InChI Formula | InChI=1 S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5 H/i1D,4 D,5 D |
| SMILES | [2 H]c1cc([2 H])c(Br)c([2 H])c1 |
| IUPAC Name | 2-bromo-1,3,5-trideuteriobenzene |
| Molecular Weight (g/mol) | 160.0264 |
| Formula Weight | 158.9763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction