missing translation for 'onlineSavingsMsg'
Learn More

Bromocyclobutane, TRC

Product Code. 30348463
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348463 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348463 Supplier Toronto Research Chemicals Supplier No. TRCB685023500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocyclobutane
CAS 4399-47-7
Molecular Formula C4H7Br
Quantity 500 mg
InChI Formula InChI=1S/C4H7Br/c5-4-2-1-3-4/h4H,1-3H2
SMILES BrC1CCC1
IUPAC Name bromocyclobutane
Molecular Weight (g/mol) 135
Formula Weight 133.9731
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.