missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocyclohexane-d11, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Bromocyclohexane-D11 |
| CAS | 35558-49-7 |
| Molecular Formula | C6 D11 Br |
| Quantity | 0.5 g |
| Synonym | Cyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11, 6-bromo- (ACI), 6-Bromocyclohexane-1,1,2,2,3,3,4,4,5,5,6-d11 (ACI), Cyclohexane-d11, bromo- (9 CI), Cyclohexyl-d11 bromide, Perdeuterated bromocyclohexane, Bromocyclohexane-d11, Bromocyclohexane-D11 |
| InChI Formula | InChI=1 S/C6H11Br/c7-6-4-2-1-3-5-6/h6H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(Br)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1-bromo-1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexane |
| Molecular Weight (g/mol) | 174.1233 |
| Formula Weight | 173.0735 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction