missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl Methyl Ether, TRC

Product Code. 30341739
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341739 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341739 Supplier Toronto Research Chemicals Supplier No. TRCB41533525G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromomethyl Methyl Ether
CAS 13057-17-5
Molecular Formula C2H5BrO
Quantity 25 g
Synonym Bromomethoxymethane, 1-Bromomethyl Methyl Ether, Methoxymethyl Bromide
InChI Formula InChI=1S/C2H5BrO/c1-4-2-3/h2H2,1H3
SMILES COCBr
IUPAC Name bromo(methoxy)methane
Molecular Weight (g/mol) 124.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.