missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromotrimethylsilane, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Bromotrimethylsilane |
| CAS | 2857-97-8 |
| Molecular Formula | C3H9BrSi |
| Quantity | 50 g |
| Synonym | NSC 139857, Trimethylbromosilane, Trimethylsilicon Bromide, Trimethylsilyl Bromide, TMS Bromide, TMSBr |
| InChI Formula | InChI=1S/C3H9BrSi/c1-5(2,3)4/h1-3H3 |
| SMILES | C[Si](C)(C)Br |
| IUPAC Name | bromo(trimethyl)silane |
| Molecular Weight (g/mol) | 153.09 |
| Formula Weight | 151.9657 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction