missing translation for 'onlineSavingsMsg'
Learn More

Bromotris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 11312669
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312669 5 g 5g
11302669 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312669 Supplier Thermo Scientific Alfa Aesar Supplier No. H26708.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50296-37-2
Molecular Formula C6H18BrF6N3P2
Molecular Weight (g/mol) 388.07
MDL Number MFCD00191864
InChI Key XELPBWPBGHCIKX-UHFFFAOYSA-N
Synonym bromotris dimethylamino phosphonium hexafluorophosphate, brop, bromotris dimethylamino phosphonium hexafluorophosphate v, bromo-tris dimethylamino phosphonium hexafluorophosphate, acmc-20aljt, ksc269e9j, bromotris dimethylamino phosphonium hexafluorophos, bromotris dimethylamino phosphoniumhexafluorophosphate, bromanyl-tris dimethylamino phosphanium hexafluorophosphate
PubChem CID 10221847
IUPAC Name bromo-tris(dimethylamino)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CN(C)[P+](Br)(N(C)C)N(C)C

Specifications

Melting Point >300°C
Quantity 5 g
UN Number UN1759
Sensitivity Light sensitive
Formula Weight 388.07
Percent Purity ≥98%
Chemical Name or Material Bromotris(dimethylamino)phosphonium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.