missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BSSP4 Monoclonal Antibody (246744)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561089
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
Specifications
| BSSP4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9ER10 | |
| PRSS22 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tryptase epsilon /BSSP-4, Ala33-Ser306 (predicted). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| 246744 | |
| Unconjugated | |
| PRSS22 | |
| 4733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO343 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 70835 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction