missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG2 Antibody (OTI4C10), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71339PE
This item is not returnable.
View return policy
Description
BTG2 Monoclonal antibody specifically detects BTG2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BTG2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI4C10 | |
| Western Blot, Immunohistochemistry | |
| B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21 | |
| Full length human recombinant protein of human BTG2(NP_006754) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 7832 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction