missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97381B
This item is not returnable.
View return policy
Description
BTG3 Polyclonal antibody specifically detects BTG3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BTG3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| Abundant in neuroepithelium area protein, ANATOB55, B-cell translocation gene 3, B-cell translocation gene10, BTG family member 3, BTG family, member 3, MGC8928, protein BTG3, Protein Tob5, tob55, TOB5TOFA | |
| Produced in rabbits immunized with E. coli-derived Human BTG3 fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10950 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction