missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BubR1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
227.00€ - 493.00€
Specifications
| Antigen | BubR1 |
|---|---|
| Dilution | Western Blot 1-10 μg/ml, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610974
|
Novus Biologicals
NBP3-18818 |
50 μg |
227.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605514
|
Novus Biologicals
NBP3-18818-250ug |
250 μg |
493.00€
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BubR1 Polyclonal antibody specifically detects BubR1 in Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| BubR1 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Sheep | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| PBS | |
| Bub1A, BUBR1budding uninhibited by benzimidazoles 1 (yeast homolog), beta, budding uninhibited by benzimidazoles 1 homolog beta (yeast), EC 2.7.11.1, hBUBR1, MAD3/BUB1-related protein kinase, MAD3LBUB1beta, Mitotic checkpoint kinase MAD3L, Protein SSK1, SSK1mitotic checkpoint serine/threonine-protein kinase BUB1 beta | |
| Recombinant protein corresponding to amino acids 232-603 of the mouse Bub1b protein. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1-10 μg/ml, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| Q9Z1S0 | |
| 701 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title