Learn More
BUCHI Boiling Point Tubes and Capillaries
Product Code. 12725063 Shop All Buchi Products
Change view
Click to view available options
Product Type:
Boiling Point Capillaries
Boiling Point Tubes
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Packaging Quantity Unit Size
12725063 Boiling Point Tubes 10/Pk. Pack of 10
12030592 Boiling Point Tubes 100/Pk. Pack of 100
12201507 Boiling Point Capillaries 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12725063 Supplier BUCHI Supplier No. 019697
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
  • High precision for reliable results
  • Meets Siwoloboff and Eu Pharmacopeia standards
  • Designed for BUCHI melting and boiling point instruments
  • Compatible with other instruments
TRUSTED_SUSTAINABILITY
Specifications
Product Type Boiling Point Tubes
Diameter (Metric) 2.75 mm
For Use With (Application) For reproducible boiling point determinations
For Use With (Equipment) Boiling point apparatus
Material Borosilicate Glass 3.3
Packaging Quantity 10/Pk.