Learn More
BUCHI Distillation Spiders for Five-Flask Distillations
Product Code. 10354443 Shop All Buchi Products
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
Taper:
29.2/32
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Taper Unit Size
10354443 50 mL 29.2/32 Each
10230262 100 mL 29.2/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10354443 Supplier BUCHI Supplier No. 001332
Estimated Shipment: 20-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Process up to 5 samples in a single parallel evaporation process
  • 5 positions
  • Holds 50 or 100mL cylindrical flasks
  • STJ 29/32 or 24/40
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Distilling Spider
For Use With (Equipment) 5 flask distillation
Taper 29.2/32
Volume (Metric) 50 mL