Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044045
Additional Details : Weight : 2.10000kg
 View more versions of this product
Product Code. 12739485
  • 1590.00€ / Each
Estimated Shipment: 13-05-2024
to see stock.
Specifications
49 mm
460 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
290 psi
Column