Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044036
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 12765995
  • 1455.00€ / Each
Estimated Shipment: 14-05-2024
to see stock.
Specifications
26 mm
230 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
580 psi
Column