Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044053
Additional Details : Weight : 5.80000kg
 View more versions of this product
Product Code. 12799485
  • 5950.00€ / Each
Estimated Shipment: 14-05-2024
to see stock.
Specifications
100 mm
460 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
145 psi
Column