Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
Product Code. 12736400 Shop All Buchi Products
Change view
Click to view available options
Product Type:
Calibration Set
IQ/OQ Protocol
Verification Kit
For Use With (Equipment):
BUCHI Melting Point Instrument
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
12736400 IQ/OQ Protocol BUCHI Melting Point Instrument Each
12053303 Calibration Set BUCHI Melting Point Instrument Each
12063303 Verification Kit BUCHI Melting Point Instrument Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12736400 Supplier BUCHI Supplier No. 11055004
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
  • Guided calibrations and verifications provide easy and reliable results
  • Ensures compliance from 10°C to 400°C
TRUSTED_SUSTAINABILITY
Specifications
Product Type IQ/OQ Protocol
For Use With (Equipment) BUCHI Melting Point Instrument
Includes Instrument logbook, IQ/OQ documentation, OQ label, calibration and verification M-565m, 100 melting point capillaries