missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bucladesine Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Bucladesine Sodium Salt |
| CAS | 16980-89-5 |
| Molecular Formula | C18H23N5NaO8P |
| Quantity | 500 mg |
| Synonym | 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, Adenosine deriv., Bucladesine Sodium, Bucladesine Sodium Salt, DC 2797, Dibutyryl 3',5'-Cyclic AMP Sodium Salt, Dibutyryl Cyclic AMP Monosodium Salt, Dibutyryl Cyclic AMP Sodium, Dibutyryl Cyclic AMP Sodium Salt, Dibutyryl-cAMP Sodium Salt, Monosodium Dibutyryl Cyclic AMP, N-6,O-2'-Dibutyryladenosine Cyclic 3',5'-Monophosphate Sodium Salt, N6,2'-O-Dibutyryl-3',5'-cyclic AMP Sodium Salt, N6,2'-O-Dibutyryladenosine-3',5'-monophosphate Monosodium Salt, N6,O2'-Dibutyryl-3',5'-cyclic AMP Sodium Salt, Sodium N6,2'-O-dibutyryladenosine 3',5'-Cyclic Phosphate, Sodium Dibutyryl 3',5'-Cyclic AMP, Sodium Dibutyryl cAMP, N-(9-β-D-Ribofuranosyl-9H-purin-6-yl)-butyramide Cyclic Hydrogen Phosphate Butyrate (ester), Monosodium Salt |
| InChI Formula | InChI=1S/C18H24N5O8P.Na/c1-3-5-11(24)22-16-13-17(20-8-19-16)23(9-21-13)18-15(30-12(25)6-4-2)14-10(29-18)7-28-32(26,27)31-14;/h8-10,14-15,18H,3-7H2,1-2H3,(H,26,27)(H,19,20,22,24);/q;+1/p-1/t10-,14-,15-,18-;/m1./s1 |
| SMILES | [Na+].CCCC(=O)Nc1ncnc2c1ncn2[C@@H]3O[C@@H]4COP(=O)([O-])O[C@H]4[C@H]3OC(=O)CCC |
| Molecular Weight (g/mol) | 491.37 |
| Formula Weight | 491.1182 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?