missing translation for 'onlineSavingsMsg'
Learn More

Bucladesine Sodium Salt, TRC

Product Code. 30375394
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375394 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375394 Supplier Toronto Research Chemicals Supplier No. TRCD49377850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bucladesine Sodium Salt
CAS 16980-89-5
Molecular Formula C18H23N5NaO8P
Quantity 50 mg
Synonym 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, Adenosine deriv., Bucladesine Sodium, Bucladesine Sodium Salt, DC 2797, Dibutyryl 3',5'-Cyclic AMP Sodium Salt, Dibutyryl Cyclic AMP Monosodium Salt, Dibutyryl Cyclic AMP Sodium, Dibutyryl Cyclic AMP Sodium Salt, Dibutyryl-cAMP Sodium Salt, Monosodium Dibutyryl Cyclic AMP, N-6,O-2'-Dibutyryladenosine Cyclic 3',5'-Monophosphate Sodium Salt, N6,2'-O-Dibutyryl-3',5'-cyclic AMP Sodium Salt, N6,2'-O-Dibutyryladenosine-3',5'-monophosphate Monosodium Salt, N6,O2'-Dibutyryl-3',5'-cyclic AMP Sodium Salt, Sodium N6,2'-O-dibutyryladenosine 3',5'-Cyclic Phosphate, Sodium Dibutyryl 3',5'-Cyclic AMP, Sodium Dibutyryl cAMP, N-(9-β-D-Ribofuranosyl-9H-purin-6-yl)-butyramide Cyclic Hydrogen Phosphate Butyrate (ester), Monosodium Salt
InChI Formula InChI=1S/C18H24N5O8P.Na/c1-3-5-11(24)22-16-13-17(20-8-19-16)23(9-21-13)18-15(30-12(25)6-4-2)14-10(29-18)7-28-32(26,27)31-14;/h8-10,14-15,18H,3-7H2,1-2H3,(H,26,27)(H,19,20,22,24);/q;+1/p-1/t10-,14-,15-,18-;/m1./s1
SMILES [Na+].CCCC(=O)Nc1ncnc2c1ncn2[C@@H]3O[C@@H]4COP(=O)([O-])O[C@H]4[C@H]3OC(=O)CCC
Molecular Weight (g/mol) 491.37
Formula Weight 491.1182
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.