Learn More
Witeg™ Buechner Filter Funnels
For use in general filtration applications in the laboratory.
Brand: Witeg™ 2 367 073
Additional Details : Weight : 0.80000kg
Description
- Made from borosilicate glass
- Multiple capacities available
- Multiple disc sizes and stem lengths
Specifications
Buechner filter funnel | |
75 mL | |
Filtration | |
P3 |
45 mm | |
Borosilicate Glass | |
10 mm |