Learn More
Invitrogen™ Dual-Light™ System Buffer A
Product Code. 12684325
Change view
Click to view available options
Quantity:
200 Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12684325 200 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12684325 Supplier Invitrogen™ Supplier No. T2176
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with Dual-Light Reporter Gene Assay System. Thermo Scientific Buffer A for Dual-Light Reporter Gene Assay System is supplied in lyophilized form.
The Dual-Light luminescent reporter gene assay is designed for the rapid and sensitive detection of firefly luciferase and E. coli β-galactosidase in a single extract aliquot for gene expression assays.
TRUSTED_SUSTAINABILITY
Specifications
Product Type β-Galactosidase/Luciferase Reporter Gene Assay System
For Use With (Equipment) Dual-Light™ Reporter Gene Assay System
Content And Storage Store the product at -20°C.
Shipping Condition Room Temperature
Product Line Dual-Light
Quantity 200 Assays
Form Solid