Learn More
Invitrogen™ Buffer Core with Cathode Bar for XCell SureLock™ Mini-Cell
Product Code. 31781211
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31781211 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31781211 Supplier Invitrogen™ Supplier No. EI9015
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Achieve fast, consistent protein gel separations in a small footprint with the updated Invitrogen XCell SureLock System. Its redesigned Buffer Core with a stainless-steel cathode bar increases run speed and system durability without compromising performance
The Invitrogen XCell SureLock Buffer Core is a key component of the XCell SureLock Mini-Cell vertical protein gel electrophoresis system. It provides a reliable electrical interface between the upper and lower buffer chambers, ensuring consistent and reproducible gel runs.
The updated Buffer Core design incorporates a stainless-steel cathode bar, platinum wire anode electrode, gold terminals, and durable silicone gaskets, offering a precise and leak-free seal during operation. This version features enhanced robustness and improved electrical efficiency for faster gel separations.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SureLock Mini
Product Line XCell
Type Electrophoresis Core
For Use With Protein Gel Electrophoresis, 1D Gel Electrophoresis, SDS PAGE, Native PAGE, IEF
Gel Type Mini
Quantity 1 Ea.