missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, Citrate Potassium Dihydrogen, pH 3.776, Fisher chemical™

Product Code. p-7083833 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12920654 500 mL Bottle 500mL
12980654 1000 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12920654 Supplier Fisher Chemical Supplier No. SOL 008 500

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifikationer

Chemical Name or Material Buffer Solution, Potassium Dihydrogen Citrate
pH 3.776 (at 25°C)
Quantity 500 mL
Packaging Bottle
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.