missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Buffered Sodium Chloride Peptone Solution (Bottled)

Product Code. 15178204
Change view
Click to view available options
Fill Weight:
100-101.5g
300 to 304 g
9.0g + 0.3g
90 to 91.5 g
Unit Size:
Pack of 10
Pack of 24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Unit Size
15178204 300 to 304 g Pack of 10
13285489 90 to 91.5 g Pack of 10
12874122 9.0g + 0.3g Pack of 24
12673418 100-101.5g Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15178204 Supplier Thermo Scientific™ Supplier No. BO0322Z
  • ONLINE EXCLUSIVE PRICE
    109.00€
    107.38€ / Pack of 10
    Save 1.62€
    1% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP/IP with this isotonic medium.

Carry out microbial limit testing by harmonized methodology of pharmaceutical products in accordance with USP/EP/BP/JP/IP with Thermo Scientific™ Buffered Sodium Chloride Peptone Solution. The combination of phosphate buffer, sodium chloride and peptone increases the viability of sensitive microorganisms by providing an isotonic environment. This is of particular importance when trying to recover cells that may be stressed or sensitive to osmotic pressure.

Use Buffered Sodium Chloride Peptone Broth for the dilution of samples in the analysis of non-sterile products for microbial contaminants.

  • Conforms to the harmonised Microbial Limit Testing chapters from the EP/USP/JP1,2,3
  • Maintains cell viability during dilution process.
  • Ready-to-use convenience of prepared media

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Content And Storage 2° to 25°C
Format 300 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 300 to 304 g
Packaging Type 10 Sirop Bottles with screw cap closure in a box
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.