Learn More
R&D Systems™ Bulk Sorting Cartridge Kit for Pala Systems (10 Cartridges)
Product Code. 31781575
Change view
Click to view available options
For Use With (Equipment):
Pala Single Cell Sorter and Dispenser
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31781575 Pala Single Cell Sorter and Dispenser 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For research use only. Handle biological samples using appropriate biosafety procedures. Cartridges are single-use and must be disposed of according to laboratory biohazard waste protocols.
Product Code. 31781575 Supplier R&D Systems™ Supplier No. NC101
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For research use only. Handle biological samples using appropriate biosafety procedures. Cartridges are single-use and must be disposed of according to laboratory biohazard waste protocols.
Previously ProteinSimple, now a part of R&D Systems. The Bulk Sorting Cartridge Kit for Pala Systems enables sterile microfluidic enrichment and collection of target cell populations. Individually packaged cartridges support efficient sorting workflows with high purity and throughput.
  • Sterile, single-use microfluidic cartridges designed for bulk sorting and enrichment of target cell populations
  • Enables efficient recovery of desired cells for downstream culture, analysis, or assay preparation
  • Supports sorting speeds up to approximately 50 events per second for high-throughput workflows
  • Microfluidic technology ensures gentle cell handling to maintain viability and functionality
  • Individually packaged cartridges reduce contamination risk and support sterile processing
  • Optimized for sample concentrations of approximately 100,000 to 500,000 cells/mL
  • Delivers enrichment purity of up to ∼90% depending on sample characteristics
  • Disposable design eliminates cleaning steps and reduces instrument downtime
  • Compatible with R&D Systems™ Pala and Hana Single Cell Dispenser systems
  • Ideal for cell population enrichment, rare cell recovery, and preparative workflows
  • Kit includes 10 cartridges for consistent and reproducible sorting performance
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cartridge
For Use With (Equipment) Pala Single Cell Sorter and Dispenser
Includes 10 Sterile Single-use Microfluidic Cartridges
Product Line ProteinSimple
Designed for use with Pala systems. Cartridges are sterile and individually packaged for single-use workflows. Sorting performance and purity depend on sample quality and preparation.