missing translation for 'onlineSavingsMsg'
Learn More

Bupropion hydrochloride, 99%, Thermo Scientific™

A selective inhibitor of dopamine uptake

Brand:  Thermo Scientific Alfa Aesar J61105.03

Quantity 1g

 View more versions of this product

Product Code. 15443759

  • 196.00€ / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.

Solubility
Soluble in water (>25 mg/ml), ethanol 193mg/ml, 0.1 N HCl 333 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

31677-93-7
276.201
HEYVINCGKDONRU-UHFFFAOYSA-N
62884
CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl
C13H19Cl2NO
MFCD00055209
bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride

Specifications

31677-93-7
MFCD00055209
14,1499
Soluble in water (>25mg/ml),ethanol 193mg/ml,0.1 N HCl 333mg/ml
CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl
276.201
276.2
Bupropion hydrochloride
C13H19Cl2NO
1g
bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
HEYVINCGKDONRU-UHFFFAOYSA-N
2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
62884
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.